Learn More
Edmund Optics OPTICAL ADHESIVE F42-192
Supplier:  Edmund Optics 42192
Encompass
Norland Optical Adhesive, 1 oz., Norland number: 60, Viscosity: 300 cps, Tough, Typical application: General purpose adhesive, Has long shelf life
Catalog No. NC9091413
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.