Learn More
Thermo Scientific™ Optical Adhesive Film, for Piko PCR plates
Catalog No. ASF0020100 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Sheets
400 Sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ASF0020100 100 Sheets
ASF0020 400 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ASF0020100 Supplier Thermo Scientific™ Supplier No. ASF0020100
Add to Cart
Add to Cart
The exceptionally clear Thermo ScientificPiko PCR/qPCR Film Seal allows high transmission of fluorescence across the range of commonly-used fluorophores.
The exceptionally clear Thermo ScientificPiko PCR/qPCR Film Seal allows high transmission of fluorescence across the range of commonly-used fluorophores. When used with aPiko Thermal CyclerorPikoReal Real-Time PCR System, sample volumes as low as 5μL can be used with minimal evaporative loss. These optical adhesive films are available in the ¼-size format for Piko PCR Plates.
When used with a Piko Thermal Cycler or PikoReal Real-Time PCR System, sample volumes as low as 5 µL can be used with minimal evaporative loss. These optical films are available in the ¼-size format for Piko PCR Plates.
Features of the Thermo Scientific Piko PCR/qPCR Film Seals
• Optical clarity with minimal auto-fluorescence for real-time PCR applications
• Tight sealing for sample volumes as low as 5 µL
• Adhesive backing allows easy application
• Cryo-compatible adhesive is effective to –80°C
Related Products
Optical Adhesive Film, for Piko PCR plates
Optical Heat Seal, for Piko PCR plates
TRUSTED_SUSTAINABILITY
Specifications
Color Optical
Quantity 100 Sheets
Product Type Film, Seal, or Applicator
Product Line Piko
Adhesive Pressure Sensitive Optical Seal
Plate Compatibility Piko Plate
Are Thermo Scientific adhesive seals free of DNase, RNase, and human DNA?
Whilst Thermo Scientific adhesive seals are manufactured in a clean facility, they are not certified free of DNase, RNase, and human DNA.
I am having problems with evaporation when using an adhesive seal. What do you recommend?
To prevent evaporation when using adhesive seals, firm application pressure must be applied to establish robust adhesive contact and bond strength around well rims. Adhesive seals such as ABsolute qPCR Seal (Cat. No. AB-1170) require sufficient pressure due to their non-tacky, pressure-sensitive adhesive. Adhesive Seal Applicator (Cat. No. AB-1391) or a similar tool may be used for optimal application of the seals. Alternatively, we recommend Thermo Scientific VersiCap Mats which show low sample evaporation with the performance of a cap. VersiCap Mats (Cat. Nos. AB-1810, AB-1815, AB-1820) offer comparable convenience and are easier to apply uniformly than adhesive seals.