Learn More
Eisco™ Optical Bench
SCI_ED
Catalog No. S23749 Shop All Eisco Products
Change view
Click to view available options
Length (English):
42 in.
Catalog No. Length (English)
S23749 42 in.
1 options
Catalog No. S23749 Supplier Eisco™ Supplier No. PH0616EMS
Add to Cart
Add to Cart
Includes
Set comprises of double concave and convex lens, plano convex and concave lens, LED, Light source, Candle holder, Lens holder, Single slit with holder, White screen, Translucent screen
Used to measure the focal length of a lens, for physics experiments.
  • Constructed with square section aluminum tube
  • Rail with 1cm graduations up to 100cm
  • With 5 riders for lens and slit holder
Specifications
Product Type Optical Bench
Description Made from square section aluminum tube;
For Use With (Application) Measuring focal length of lens
Includes 5 riders for lens and slit holder
Length (English) 42 in.