Learn More
United Scientific™ Optical Bench Set With Meter Stick
SDPSCI_ED
Catalog No. S05912 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S05912 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. S05912 Supplier United Scientific™ Supplier No. OBSET1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Optical Bench Set With Meter Stick contains sufficient equipment for high school physics applications.
Contents:
  • 1 Meter Stick
  • 1 Screen support
  • 2 Lens Supports for 38 mm and 75 mm lenses/mirrors
  • 5 Board screens measuring 10 x 12.5 cm with a millimeter scale on both sides
  • 1 Marker and object
  • 2 Metal supports for meter stick
  • 1 Candle holder
  • 1 Paraffin candle
Note: Replacement meter stick available (catalog number S05922)
Specifications
Quantity 1 Ea.
Material Wood
Includes Meter Stick
For Use With (Application) High school physics applications
Product Type Optical Bench Set
Shape Various