Learn More
Vernier™ Optical DO Probe Metal Guard
SDPSCI_ED
Catalog No. S16230ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Optical DO Probe
Catalog No. For Use With (Equipment)
S16230ND Optical DO Probe
1 options
Catalog No. S16230ND Supplier Vernier™ Supplier No. ODOGRD
Add to Cart
Add to Cart
Includes
Optical DO Probe Metal Guard
The Optical DO Probe Metal Guard protects the Vernier Optical DO Probe cap when taking measurements in the field and helps weigh down the probe when submerged.
  • Protects the Vernier™ Optical DO Probe cap when taking measurements
  • Helps weigh down the probe when submerged
Warranty and Services
5-year limited warranty
Specifications
Material Metal
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Optical DO Probe
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov