Learn More
Vernier™ Optical DO Probe Replacement Cap
SDPSCI_ED
Catalog No. S16229ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Optical DO Probe
Catalog No. For Use With (Equipment)
S16229ND Optical DO Probe
1 options
Catalog No. S16229ND Supplier Vernier™ Supplier No. ODOCAP
Add to Cart
Add to Cart
Includes
Optical DO Probe Replacement Cap
This is a replacement cap for the Vernier Optical DO Probe. The cap will need to be replaced every few years. Each cap is supplied with a unique calibration code on a microSD card.
  • This is a replacement cap for the Vernier™ Optical DO Probe.
  • The cap will need to be replaced every few years.
  • Each cap is supplied with a unique calibration code on a microSD card.
Warranty and Services
2-year limited warranty
Specifications
Type Cap
Includes Calibration code on a microSD card
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Optical DO Probe