Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Vernier Optical DO Probe
SDPSCI_ED

Catalog No. S16228ND Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 20 mg/L (or ppm), 0 to 300%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S16228ND 0 to 20 mg/L (or ppm), 0 to 300%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16228ND Supplier Vernier™ Supplier No. ODOBTA

Includes

Vernier Optical DO Probe

Requires

Requires an interface and software, sold separately

The Vernier Optical DO Probe makes it easy to measure the dissolved oxygen concentration in water. Perfect for the field or for the laboratory, this plug-and-play probe requires no calibration, no filling solution, no warm-up time, and no stirring.

Range: 0 to 20 mg/L (or ppm), 0 to 300% Accuracy: mg/L ±0.2 mg/L below 10 mg/L ±0.4 mg/L above 10 mg/L % ±2% below 100% ±5% above 100% Response Time: 90% of final reading in 40 seconds Typical Resolution: 0.006 mg/L Temperature Compensation: automatic from 0–50°C Pressure Compensation: automatic from 228 mmHg to 1519 mmHg Minimum sample flow: none

Warranty and Services

5 years (2 years for cap)

Specifications

Grade High School and College
Interface Requires
Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy ±0.2 mg/L below 10 mg/L, ±0.4 mg/L below 10 mg/L
Range 0 to 20 mg/L (or ppm), 0 to 300%
Resolution 0.006 mg/L
Response Time 90% of final reading in 40 seconds
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.