Learn More
NEWPORT FRANKLIN INC OPTICAL FILTER, 10MLF10-405
Supplier:  NEWPORT FRANKLIN INC 10MLF10405
Encompass
Optical Filter, Mercury Line, 25.4 mm, 404.7±2 nm Center 10±2nm FWHM
Catalog No. NC2084222
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.