Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Opticap™ Sterile XL300 Capsule Filters

Catalog No. KHGES003HH3 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.5/0.1 μm
0.5/0.2 μm
Inlet Style:
0.56 in. (14 mm) Hose Barb
0.56 in. (14mm) Hose Barb
0.75 in. (19mm)Sanitary Flange
Outlet Style:
0.56 in. (14mm) Hose Barb
0.75 in. (19mm)Sanitary Flange
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Inlet Style Outlet Style
KHGES003HH3 0.5/0.2 μm 0.56 in. (14mm) Hose Barb 0.56 in. (14mm) Hose Barb
KHVES003FF3 0.5/0.1 μm 0.75 in. (19mm)Sanitary Flange 0.75 in. (19mm)Sanitary Flange
KHVES003FH3 0.5/0.1 μm 0.75 in. (19mm)Sanitary Flange 0.56 in. (14mm) Hose Barb
KHGES003FF3 0.5/0.2 μm 0.75 in. (19mm)Sanitary Flange 0.75 in. (19mm)Sanitary Flange
KHGES003FH3 0.5/0.2 μm 0.75 in. (19mm)Sanitary Flange 0.56 in. (14mm) Hose Barb
KHVES003HH3 0.5/0.1 μm 0.56 in. (14 mm) Hose Barb 0.56 in. (14mm) Hose Barb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. KHGES003HH3 Supplier MilliporeSigma™ Supplier No. KHGES003HH3
Add to Cart
Add to Cart
This item is not returnable. View return policy

For sterilizing grade performance and mycoplasma removal

  • High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
  • On-board PES membrane prefilter for extended throughput in high fouling media
  • Robust construction
  • Withstand multiple sterilization cycles
  • Easy wetting and integrity testing
  • Broad chemical compatibility across a wide pH range (pH 1–14)
  • Hydrophilic Polyethersulfone filter
  • Polyester support

  • Structural Components: Gamma stable polypropylene/Polysulfone
  • Silicone vent O-rings
  • Gamma compatible sterilization

Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals

TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PES
Pore Size 0.5/0.2 μm
Sterility Sterile
Filtration Area 0.03 m2
For Use With (Application) Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Inlet Style 0.56 in. (14mm) Hose Barb
Outlet Style 0.56 in. (14mm) Hose Barb
Length (English) 4.7 in.
Length (Metric) 11.9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.