Learn More
MilliporeSigma Opticap XL10 Durapore
Supplier:  MilliporeSigma KHGLA10HH1
Encompass
Sterile, inlet connection diam. 9/16 in., pore size 0.45/0.22 μm, cartridge nominal length 10 in. (25 cm).
Catalog No. NC1361137
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.