Learn More
Chemglass Life Sciences OptiChem Heating Mantle Controller, Single Circuit, 10A, 1200 Watts, 120VAC
SDP
Catalog No. 50401788
Encompass_Preferred
OptiChem Heating Mantle Controller, Single Circuit, 10A, 1200 Watts, 120VAC. Our controllers feature solid-state proportional voltage power control in a compact benchtop design. Controllers provide accurate manual control of heating mantles, tapes and all other resistive loads with an adjustable output range from 5 to 100% of the rated voltage. All units are supplied with a 6ft separable power cord, a grounded, 3-wire receptacle and lighted power switch. 3-Year warranty.
Catalog No. 50-401-788 Supplier Chemglass Life Sciences Supplier No. CG1500501
OptiChem Heating Mantle Controller, Single Circuit, 10A, 1200 Watts, 120VAC. Our controllers feature solid-state proportional voltage power control in a compact benchtop design. Controllers provide accurate manual control of heating mantles, tapes and all other resistive loads with an adjustable output range from 5 to 100% of the rated voltage. All units are supplied with a 6ft separable power cord, a grounded, 3-wire receptacle and lighted power switch. 3-Year warranty.