Learn More
InVitria Optiferrin - Recombinant Human Transferrin - 10g
Supplier: InVitria 777TRF02910G
InVitria’s Optiferrin recombinant human transferrin is completely animal- and blood-free. Its functional performance in cell culture is equivalent to human holo-transferrin in terms of reversible iron binding, transferrin-receptor binding, and promoting cell growth and productivity. It is compatible with a variety of cell types and applications including stem, primary, bioproduction cells, proliferation of human HSC, and Sp2/0 hybridoma cells. Optiferrin offers significant regulatory advantages due to its safety and consistency.Made in the USA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.