Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Optifit Extended Plus Tip
GREENER_CHOICE

Catalog No. 12133945 Shop All Sartorius Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
12-133-945 Sterile
12-133-944 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-133-945 Supplier Sartorius Supplier No. LHX791001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware.

  • The Optifit Extended Plus tips are designed to improve pipetting reach, making them ideal for use with deep and narrow labware
  • The tip design allows for more efficient retrieval of samples, down to the last drop
  • The longer length of these tips helps prevent contamination by ensuring that the pipette does not come into contact with the walls of labware or containers during use
  • The tips are manufactured in an ISO 8 certified cleanroom environment and are certified to be free of DNase, RNase, endotoxin and human DNA
  • Sartorius pipettes and tips are designed to seamless fit together for the most reliable and reproducible results
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 mL
Filtered Non-filtered
Sterility Sterile
Color Transparent
Autoclavable Autoclavable
Format Rack
Tip Style Ergonomic
Product Type Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.