Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Optifit tip, 1000μL, Wide Bore, Flexibulk (480)
GREENER_CHOICE

Catalog No. 14577817 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-577-817 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-577-817 Supplier Sartorius Supplier No. LHB791024
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optifit tips ensure perfect fitting and leak-tight sealing on Sartorius pipettes for maximum accuracy and reproducibility. These nonsterile 10–1,000μL wide bore tips are manufactured in an ISO 8 certified cleanroom environment and are DNase-, RNase- and endotoxin-free. Provided in an economical bulk box.

These Non-filtered Optifit Tips enable ergonomic and light tip attachment by being compatible with the Optiload feature in Sartorius Picus™ NxT, Picus™, eLINE™, mLINE™ and Proline™ Plus pipettes. Optifit Tips are available in Flexibulk packaging, a space-saving, ecological and economical bulk box (480 tips).

  • Strict quality standards, ISO 9001 and ISO 13485, are followed from R and D to production and delivery
  • Manufactured in ISO 8 classified clean room conditions to avoid any contaminations
  • Tested for DNase, RNase and endotoxins per each tip lot
  • Wide bore tips measure 68.5mm long with a volume of 10-1,000μL
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Transparent
Autoclavable Autoclavable
Length (Metric) 68.5mm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) Precision Liquid Dispensing
Format Bulk
Tip Style Genomic/Wide Orifice
Product Type Non-filtered Tips
Disposable Single-use
For Use With (Equipment) Sartorius Biohit Picus, Sartorius eLINE, Sartorius mLINE, Sartorius Proline Plus
Graduated No
Product Line Optifit
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.