Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Optifit Tip
GREENER_CHOICE

Catalog No. 14558232 Shop All Sartorius Products
Change view
Click to view available options
Format:
Bulk
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14-558-232 Bulk
14-558-231 Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-558-232 Supplier Sartorius Supplier No. LH799004
Add to Cart
Add to Cart

Optifit tips are ideal for use with for Sartorius pipettes, enabling maximum accuracy and precision, as well as comfortable ergonomics.

  • Non-filtered Optifit tips are designed to seamlessly fit together with Sartorius pipettes (Picus 2, Picus, Picus NxT, Tacta, Mline, Proline Plus).
  • Feature a unique spring-loaded Optiload mechanism that makes it easy to attach and eject tips while still ensuring a perfect seal.
  • Available in various packaging types, purity certified and pre-sterilized.
  • For applications involving larger volume transfers, the 10 mL Optifit tips are specifically designed to deliver precise and accurate performance.
  • Tips are manufactured in an ISO 8 certified cleanroom environment.
  • Components, including the tips, trays, and racks, are made from fully recyclable polypropylene.
  • Racks and tips can be autoclaved (121ºC for 20 minutes).
  • Note: For Waters™ Andrew+™ Pipetting Robot, please visit Waters e-shop for compatible 10 mL tips (SKU: 186010204).
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10,000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Transparent
Autoclavable Not Autoclavable
Format Bulk
Tip Style Ergonomic
For Use With (Equipment) Sartorius
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.