Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles

Catalog No. 09981036
Change view
Click to view available options
Diameter (Metric):
0.2 μm
0.3 μm
0.4 μm
0.5 μm
0.85 μm
2 μm
3 μm
4.0 μm
5.0 μm
Quantity:
15 mL
100 mL
Surface Acid Loading:
High Acid
Low Acid
Medium Acid
Concentration:
10%
4%
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity Surface Acid Loading Concentration
09-981-036 0.2 μm 15 mL High Acid 10%
09-981-057 2 μm 15 mL High Acid 10%
09-981-058 2 μm 100 mL High Acid 10%
09-981-060 3 μm 15 mL High Acid 10%
09-981-061 3 μm 100 mL High Acid 10%
09-981-037 0.2 μm 100 mL High Acid 10%
09-981-033 0.2 μm 15 mL Medium Acid 10%
09-981-034 0.2 μm 100 mL Medium Acid 10%
09-981-045 0.3 μm 15 mL High Acid 10%
09-981-046 0.3 μm 100 mL High Acid 10%
09-981-044 0.3 μm 15 mL Medium Acid 10%
09-981-043 0.3 μm 100 mL Medium Acid 10%
09-981-054 0.4 μm 15 mL High Acid 10%
09-981-055 0.4 μm 100 mL High Acid 10%
09-981-049 0.4 μm 100 mL Low Acid 10%
09-980-862 0.2 μm 15 mL Low Acid 10%
09-980-863 0.2 μm 100 mL Low Acid 10%
09-980-690 4.0 μm 15 mL High Acid 4%
09-980-691 4.0 μm 100 mL High Acid 4%
09-980-692 5.0 μm 15 mL High Acid 4%
09-980-693 5.0 μm 100 mL High Acid 4%
09-980-684 0.5 μm 15 mL Medium Acid 4%
09-980-686 0.85 μm 15 mL Medium Acid 4%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 09-981-036 Supplier Thermo Scientific™ Supplier No. 83000520100290
Add to Cart
Add to Cart

Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.

Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.

Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization. The surface area per gram of a particle varies inversely with the particle diameter. Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based.
  • For most applications, Opti-Link particles can be used directly from the bottle without any pre-cleaning
  • Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces, nor cause proteins to desorb from particle surfaces
  • Various surface acid concentrations available to help optimize reagent development efforts
  • Opti-Link particles have been used in and are equally successful in molecular biology and research applications

Parking Area Properties

  • Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
  • Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
  • High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling

Recommended for:

Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays

Note:

When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.

TRUSTED_SUSTAINABILITY

Specifications

Additive Proprietary anionic surfactant
Certifications/Compliance Includes Certificate of Analysis
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
Color White or Colorless
For Use With (Application) Covalent Coupling
Material Polystyrene Sulfate
Product Type Specialty Particles
Includes Package Insert Sheet containing Certicate of Analysis
Packaging 15 mL
Particle Density 1.05g/cm³
Preservative Pasteurized
Quantity 15 mL
Sample Evaluation Packs Packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles.
Specialty Packaging Opti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request.
Surface Acid Loading High Acid
Diameter (Metric) 0.2 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.