Learn More
Beckman Coulter OptiLyse B, No-Wash Lysing Solution, 250 Tests, RUO
Supplier: Beckman Coulter IM1400
OptiLyse B is an erythrolytic reagent for lysing human red blood cells following direct immunofluorescence staining of whole blood for flow cytometry. OptiLyse B is a ready-to-use lysing solution optimized for use on BD Biosciences instruments.