Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA OptimAir™ TL PAPR Standard Kit
Standard kits come complete with PAPR, battery, charger, and appropriate breathing tube
$2787.58 - $3002.01
Specifications
Certifications (NIOSH) | NIOSH |
---|---|
For Use With (Application) | Chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical |
Includes | Motor blower, smart charger, standard battery, hose and decon belt |
Intrinsic Safety (IS) Rating | No |
Catalog Number | Mfr. No. | Facepiece/Headtop Model | Battery Type | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Facepiece/Headtop Model | Battery Type | Product Type | Price | Quantity & Availability | |||
18-808-761
|
MSA
10081115 |
Facepiece | Lithium-ion | PAPR Assembly with Facepiece |
Each for $3,002.01
|
|
|||
18-808-763
|
MSA
10081117 |
Hood | Lithium-ion | PAPR Assembly with Hood |
Each for $2,787.58
|
|
|||
Description
- Available with facepiece or hood
- Motor blower electronics allow for optimum airflow
- Compact and ergonomic battery pack available in standard or extended-life
- Automatic flow rates
- Crushproof breathing tube and sealing gaskets on cartridges
- Audible and visual alarms for low flow indication and low battery voltage
- Suitable applications include chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical
Specifications
NIOSH | |
Chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical | |
Motor blower, smart charger, standard battery, hose and decon belt | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title