Learn More
Med Vet International Optixcare, Eye EMS, (Extracellular Matrix Substitute), 5 Count
Supplier: Med Vet International PHOPTIXCAREEMS
Optixcare Eye EMS is indicated to help promote rapid healing, comfort and recovery in pets suffering from acute or chronic non-healing corneal ulcers. It contains a RGTA exclusive regenerative technology that quickly promotes corneal healing by providing an extra-cellular matrix substitute that is resistant to degradation.-For dogs, cats, horses and small mammals-Monodose for hygienic precise dosing-Preservative freeProducts are not always manufacturer specific. Actual product appearance may differ from website image.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.