Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ AloI, Fisher BioReagents™

Catalog No. BP80751 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP8075-1 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP8075-1

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

5'...^ 7(N) G A A C (N)6 T C C (N)12-13^...3'
3'...^12-13(N) C T T G (N)6 A G G (N)7 ^...5'

Supplied with: 10X OPTIZYME Buffer 5
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 30°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: 0 - 20%
  • Buffer 3: 0 - 20%
  • Buffer 4: 20 - 50%
  • Buffer 5: 100%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • Approximately 70% of the DNA fragments can be ligated and more than 80% of these can be re-cut after 10-fold over-digestion with AloI.
  • Notes:
    • Incubation at 37°C results in 20% activity.
    • AloI produces double-strand cuts on both sides from the interrupted recognition site. Its unique feature is a degenerate cleavage point on the 3 side of the recognition sequence (12 or 13 nucleotides away).
    • The presence of S-adenosylmethionine in the reaction mixture results in incomplete cleavage with AloI.
    • Greater than 10-fold overdigestion with AloI may result in star activity.
    • AloI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.
    Methylation Effects:
    Dam: May overlap - effect not determined
    • 5'...GAAC(N)4Gm6A TCC...3'
  • 3'...CTTG(N)4C Tm6AGG...5'
  • (Cleavage effect not determined)
  • Dcm: May overlap - no effect
    • 5'...GAAC(N)6TCm5CW GG...3'
  • 3'...CTTG(N)6AG GWm5CC...5'
  • (No effect on cleavage)
  • CpG: May overlap - cleavage impaired
    • 5'...m5C GAAC(N)6TCC...3'
  • 3'... Gm5CTTG(N)6AGG...5'
  • (Cleavage is blocked)
    • 5'...GAAC(N)6TCm5C G...3'
  • 3'...CTTG(N)6AG Gm5C...5'
  • (No effect on cleavage)
    • 5'...GAAm5C G(N)5TCC...3'
  • 3'...CTT Gm5C(N)5AGG...5'
  • (No effect on cleavage)
  • EcoKI: Never overlaps - no effect
  • EcoBI: Never overlaps - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 1 to 3 U/μL
    Components 25 to 50% Water, >50% Glycerin
    Incubator Temperature 30°C
    pH 7.4
    For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with AloI
    Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 100 U
    Cut Site (7/12)GAAC(N)6TCC(12/7)
    Product Type AloI
    Form Liquid
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.