Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ DNase I (RNase-Free), Fisher BioReagents™

Catalog No. BP81071 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP81071 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP81071

missing translation for 'validMainframeMsg'

OPTIZYME™ DNase I is a recombinant endonuclease that digests DNA and is RNase-Free.

Applications and Features:

  • OPTIZYME DNase I, RNase-Free is used for the preparation of DNA-free RNA
  • Removal of template DNA following in vitro transcription
  • DNA labeling by nick translation in conjunction with DNA Polymerase I
  • Preparation of DNA-free RNA prior to RT-PCR
  • Studies of DNA-protein interactions by DNase I, RNase-free footprinting
  • Enzyme activity is strictly dependent on Ca2+ and is activated by Mg2+or Mn2+ions
  • Purified from non-animal host with a lower level of intrinsic RNases

Supplied with:

  • 10X OPTIZYME DNase I Buffer: 100mM Tris-HCl (pH 7.5 at 25°C), 25mM MgCl2, 1mM CaCl2
  • 50mM OPTIZYME EDTA (for inactivation by heat at 65°C for 10min)
Source: E. Coli cells with a cloned gene encoding bovine DNase I
Quality Control:
  • Ribonuclease-free
  • Functionally tested for the digestion of template DNA after in vitro transcription

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 U/μL
Color Colorless
pH 7.5
For Use With (Application) Preparation of DNA-free RNA, removal of template DNA following in-vitro transcription, DNA labeling by nick translation in conjunction with DNA Polymerase I, Preparation of DNA-free RNA prior to RT-PCR, Studies of DNA-Protein interactions by DNase I, RNase-free footprinting
Storage Buffer OPTIZYME™ DNase I Buffer 10X : Tris HCl 100mM (7.5 pH at 25°C), MgCl2 25mM, CaCl2 1mM, OPTIZYME™ EDTA 50mM (for inactivation by heat at 65°C for 10min.)
Content And Storage -20°C
Source E. coli cells with a cloned gene encoding bovine DNase I
Quantity 1000 U
Product Type DNase I (RNase-free)
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.