Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ RsaI, Fisher BioReagents™

Catalog No. bp80001 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP8000-1 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP8000-1

5'...G T^A C...3'
3'...C A^T G...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 50 - 100%
  • Buffer 2: 20 - 50%
  • Buffer 3: 0 - 20%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
    • More than 95% of the DNA fragments can be ligated and re-cut after a 50-fold over-digestion with RsaI
  • Methylation Effects:
  • Dam: Never overlaps - no effect
  • Dcm: Never overlaps - no effect
  • CpG: May overlap - cleavage impaired
    • 5'...GTAm5C G...3'
  • 3'...CAT Gm5C...5'
  • (Cleavage is impaired)
    • 5'...m5C GTAm5C G...3'
  • 3'... Gm5CAT Gm5C...5'
  • (Cleavage is blocked)
  • EcoKI: Never overlaps - no effect
  • EcoBI: Never overlaps - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components >50% Glycerin, 25 to 50% Water
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 10-fold over-digestion with Rsa I
    Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 1000 U
    Cut Site GT.AC
    Product Type RsaI
    Form Liquid
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.