Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ T3 RNA Polymerase, Fisher BioReagents™

Catalog No. bp81011 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP81011 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP81011

OPTIZYME™ T3 RNA Pol is a DNA-dependent RNA polymerase that catalyzes the 5'->3' synthesis of RNA from DNA.

Applications and Features:

  • OPTIZYME™ T3 RNA Polymerase synthesizes unlabeled and labeled RNA that can be used for: (1) hybridization, (2) in vitro RNA translation, (3) RNase protection assays, (4) genomic DNA sequencing, (5) RNA splicing, and (6) studies of RNA secondary structure and RNA-protein interactions
  • Incorporation of modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)

Supplied with:
5X OPTIZYME Transcription Buffer: 200mM Tris-HCl (pH 7.9 at 25°C), 30mM MgCl2, 50mM DTT, 50mM NaCl, 10mM spermidine
Source: E. Coli cells with a cloned T3 RNA Polymerase gene
Quality control:

  • Endo-, exodeoxyribonuclease and ribonuclease-free
  • Functionally tested in in vitro transcription reaction

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/μL
Color Colorless
pH 7.5
For Use With (Application) Hybridization, in-vitro RNA translation, RNase protection assay, genomic DNA sequencing, RNA splicing and studies of RNA secondary structure and RNA-Protein interaction, quality control
Storage Buffer OPTIZYME™ Transcription Buffer 5X : Tris HCl 200mM (7.9 pH at 25°C), MgCl2 30mM, DTT 50mM, NaCl 50mM, Spermidine 10mM
Content And Storage -20°C
Source E. coli cells with a cloned T3 RNA Polymerase gene
Quantity 1000 U
Product Type T3 RNA Polymerase
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.