Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ T4 DNA Polymerase, Fisher BioReagents™

Catalog No. BP81051 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP81051 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP81051

missing translation for 'validMainframeMsg'

OPTIZYME™ T4 DNA Pol is a template-dependent DNA polymerase that catalyzes 5'->3' synthesis from primed single-stranded DNA.

Applications and features:

  • OPTIZYME T4 DNA Polymerase creates blunt DNA ends by filling in 5'-overhangs and/or removing 3'-overhangs
  • Creation of blunt PCR products with 3'-dA overhangs
  • Oligonucleotide-directed site-specific mutagenesis
  • Synthesis of labeled DNA probes (by replacement reaction)
  • Ligation-independent cloning of PCR products
  • Active in buffers for OPTIZYME restriction enzymes, reverse transcriptases and T4 DNA ligase
  • 3'->5' exonuclease activity
  • Lacks 5'->3' exonuclease activity

Supplied with:
5X OPTIZYME T4 DNA Pol Buffer: 335mM Tris-HCl (pH 8.8 at 25°C), 33mM MgCl2, 5mM DTT, 84mM (NH4)2SO4

Source: E. Coli cells with a cloned gene 43 of bacteriophage T4

Quality Control:
Endodeoxyribonuclease-free

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Color Colorless
pH 7.5
For Use With (Application) Synthesis of labeled DNA probes, creation of blunt PCR products with 3'-dA overhangs, ligation-independent cloning of PCR products, Restriction Enzymes, reverse transcriptases and T4 DNA ligase
Storage Buffer OPTIZYME™ T4 DNA Pol Buffer 5X: Tris HCl 335mM (8.8 pH at 25°C), MgCl2 33mM, DTT 5mM, (NH4)2SO4 84mM
Content And Storage -20°C
Source E. coli cells with a cloned gene 43 of bacteriophage T4
Quantity 100 U
Product Type T4 DNA Polymerase
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.