Learn More
Abcam Optovin, Photoactive TRPA1 Activator, 25 mg
Description
- Activity: Reversible photoactive TRPA1 activator
- Biological effect: Induces light-activated nociceptive motor behavior responses in vivo (EC50 = 2 μM)
- Molecular weight: 315.4 Da
- Storage buffer: 100% Optovin
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Format | Vial |
| Content And Storage | Store at -20°C. |
| Buffer | 100% Optovin |
| Form | Solid |
| Target Molecule | TRPA1 |
| Formulation | 100% Optovin |
| Quantity | 25 mg |
| For Use With (Application) | TRPA1 activation, nociceptive motor behavior studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.