Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Optovin, Photoactive TRPA1 Activator, 315.4 Da, 5 mg

Catalog No. 502704857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4857 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4857 Supplier Abcam Supplier No. AB1461925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reversible photoactive TRPA1 activator that induces light-activated nociceptive motor behavior responses in vivo.

  • Reversible photoactive TRPA1 activator
  • Induces light-activated nociceptive motor behavior responses in vivo (EC50 = 2 μM)
  • Molecular Weight: 315.4 Da

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule TRPA1
Content And Storage Store at -20°C.
Formulation 100% Optovin
Quantity 5 mg
For Use With (Application) TRPA1 activation, In vivo nociceptive motor behavior research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.