Learn More
TARGETMOL CHEMICALS INC OR-1896 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T123155MG
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. OR-1896 is the active metabolite of Levosimendan a highly specific phosphodiesterase (PDE III) inhibitor a vasodilator with partial anti-inflammatory properties that produces vasodilation in different types of blood vessels through activation of ATP-sensitive (KATP) and other potassium channels.OR-1896 can be used to study heart failure and vascular dysfunction. OR-1896 can be used to study heart failure and vascular dysfunction. purity: 99%
Catalog No. 50-257-7428
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.