Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ OR Grade Heath Suture Scissors - Anti-glare

Catalog No. 21124962 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
21-124-962 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-962 Supplier Surgical Design™ Supplier No. ORG138SHT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O.R.-grade Heath suture scissors with anti-glare finish provide controlled cutting for suture removal and minor procedures with dependable handling.

  • Length: 6 in.
  • Specialized Heath-style blade for easy, precise cutting of sutures
  • Angled cutting profile improves visibility and access during removal
  • Made from premium surgical stainless steel for strength and longevity
  • Anti-glare satin finish reduces reflections under operating lights
  • Autoclavable and reusable, ideal for repeated clinical use
  • Suitable for operating rooms, outpatient facilities, ER, and procedure trays

Specifications

Product Type Suture Scissors
Material Stainless Steel
Length (English) 6 in.
Length (Metric) 15.25 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.