Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR11H1/OR11H2/OR11H12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102272 Supplier Invitrogen™ Supplier No. PA5102272
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR11H1/11H2/11H12.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR11H1/OR11H2/OR11H12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR11H12
Gene Accession No. B2RN74, Q8NG94, Q8NH07
Gene Alias C14orf15; Olfactory receptor 11H1; olfactory receptor 11H12; Olfactory receptor 11H2; olfactory receptor family 11 subfamily H member 1; olfactory receptor family 11 subfamily H member 12; olfactory receptor family 11 subfamily H member 2; olfactory receptor OR14-1; Olfactory receptor OR22-1; olfactory receptor, family 11, subfamily H, member 1; olfactory receptor, family 11, subfamily H, member 12; OR11H1; OR11H12; OR11H13; OR11H2; OR11H2P; OR11H8P; OR22-1; seven transmembrane helix receptor
Gene Symbols OR11H1, OR11H12, OR11H2
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR11H1(Accession Q8NG94), corresponding to amino acid residues Q286-I326.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440153, 79334, 81061
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.