Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2A5/OR2A14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103226
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103226 100 μL
1 options

Catalog No. PIPA5103226

Supplier: Invitrogen™ PA5103226

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR2A5/2A14.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2A5/OR2A14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR2A14
Gene Accession No. Q96R47, Q96R48
Gene Alias olfactory receptor 2A14; Olfactory receptor 2A26; olfactory receptor 2A5; Olfactory receptor 2A6; olfactory receptor 2A8; Olfactory receptor 7-138/7-141; olfactory receptor family 2 subfamily A member 14; olfactory receptor family 2 subfamily A member 5; olfactory receptor OR7-12; olfactory receptor, family 2, subfamily A, member 11 pseudogene; olfactory receptor, family 2, subfamily A, member 14 pseudogene; olfactory receptor, family 2, subfamily A, member 26; olfactory receptor, family 2, subfamily A, member 5; olfactory receptor, family 2, subfamily A, member 6; olfactory receptor, family 2, subfamily A, member 8; OR2A11P; OR2A14; OR2A14P; OR2A26; OR2A5; OR2A6; OR2A8; OR7-138; OR7-141; OST182
Gene Symbols OR2A14, OR2A5
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR2A14(Accession Q96R47), corresponding to amino acid residues S242-R292.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135941, 393046
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.