Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2B6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549288
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549288 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549288 Supplier Invitrogen™ Supplier No. PA549288
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR2B6 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2B6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR2B6
Gene Accession No. P58173
Gene Alias dJ408B20.2; Hs6M1-32; olfactory receptor 2B1; Olfactory receptor 2B5; olfactory receptor 2B6; Olfactory receptor 5-40; olfactory receptor 6-31; olfactory receptor family 2 subfamily B member 6; olfactory receptor OR6-4; olfactory receptor, family 2, subfamily B, member 1 pseudogene; olfactory receptor, family 2, subfamily B, member 5; olfactory receptor, family 2, subfamily B, member 6 pseudogene; OR2B1; OR2B1P; OR2B5; OR2B6; OR2B6P; OR5-40; OR5-41; OR6-31
Gene Symbols OR2B6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 208-236 amino acids from the C-terminal region of human OR2B6.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26212
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.