Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2L13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548154
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548154 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548154 Supplier Invitrogen™ Supplier No. PA548154
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR2L13 Polyclonal Antibody for Flow, Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2L13
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR2L13
Gene Accession No. Q8N349
Gene Alias Olfactory receptor 2L13; olfactory receptor 2L14; olfactory receptor family 2 subfamily L member 13; olfactory receptor, family 2, subfamily L, member 13; olfactory receptor, family 2, subfamily L, member 14; OR2L13; OR2L14
Gene Symbols OR2L13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 244-271 amino acids from the C-terminal region of human OR2L13.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284521
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.