Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR51L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100965 Supplier Invitrogen™ Supplier No. PA5100965
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR51L1.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR51L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR51L1
Gene Accession No. Q8NGJ5
Gene Alias Olfactory receptor 51L1; olfactory receptor family 51 subfamily L member 1; olfactory receptor OR11-31; olfactory receptor, family 51, subfamily L, member 1; OR11-31; OR51L1
Gene Symbols OR51L1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR51L1(Accession Q8NGJ5), corresponding to amino acid residues V45-Q95.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119682
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.