Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR51V1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570839 Supplier Invitrogen™ Supplier No. PA570839
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

OR51V1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR51V1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR51V1
Gene Accession No. Q9H2C8
Gene Alias Odorant receptor HOR3'beta1; olfactory receptor 51A12; Olfactory receptor 51V1; olfactory receptor family 51 subfamily V member 1; olfactory receptor OR11-36; olfactory receptor, family 51, subfamily V, member 1; OR11-36; OR51A12; OR51V1
Gene Symbols OR51V1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human OR51V1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283111
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.