Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR52B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102133 Supplier Invitrogen™ Supplier No. PA5102133
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR52B2.

OR52B2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR52B2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR52B2
Gene Accession No. Q96RD2
Gene Alias Olfactory receptor 52B2; olfactory receptor family 52 subfamily B member 2; olfactory receptor OR11-70; olfactory receptor, family 52, subfamily B, member 2; OR11-70; OR52B2
Gene Symbols OR52B2
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR52B2(Accession Q96RD2), corresponding to amino acid residues M208-F258.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255725
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.