Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR5D14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571191 Supplier Invitrogen™ Supplier No. PA571191
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

OR5D14 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5D14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR5D14
Gene Accession No. Q8NGL3
Gene Alias Olfactory receptor 5D14; olfactory receptor family 5 subfamily D member 14; olfactory receptor OR11-141; Olfactory receptor OR11-150; olfactory receptor, family 5, subfamily D, member 14; OR11-141; OR11-150; OR5D14
Gene Symbols OR5D14
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human OR5D14.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219436
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.