Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR5K2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144370 Supplier Invitrogen™ Supplier No. PA5144370
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR5K2 Polyclonal Antibody for Western Blot, ELISA

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5K2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene OR5K2
Gene Accession No. Q8NHB8
Gene Alias Olfactory receptor 5K2; olfactory receptor family 5 subfamily K member 2; olfactory receptor OR3-9; olfactory receptor, family 5, subfamily K, member 2; OR3-9; OR5K2
Gene Symbols OR5K2
Host Species Rabbit
Immunogen Peptide sequence from Human Olfactory receptor 5K2 protein (6-24AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402135
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.