Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORAI1 Monoclonal Antibody (3F6H5)
GREENER_CHOICE

Catalog No. PIMA515776 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515776 Supplier Invitrogen™ Supplier No. MA515776
  • $704.00
    $550.65 / Each of 1

    Save $153.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

A suggested positive control is human ovary tissue lysate.

ORAI1 is a plasma membrane protein essential for store-operated calcium entry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORAI1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F6H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Orai1
Gene Accession No. Q5M848, Q8BWG9, Q96D31
Gene Alias calcium release-activated calcium channel 1; calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; CRACM1; D730049H07Rik; FLJ14466; hypothetical protein FLJ14466; hypothetical protein LOC402892; im:7146264; IMD9; ORAI calcium release-activated calcium modulator 1; ORAI calcium release-activated calcium modulator 1b; orai1; orai-1; orai1b; ORAT1; protein orai-1; RGD1311873; TAM2; TMEM142A; Transmembrane protein 142A; zFLJ14466; zgc:109721
Gene Symbols Orai1
Host Species Mouse
Immunogen A 16 amino acid peptide from near the carboxy terminus of human ORAI1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109305, 304496, 84876
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.