Learn More
AGI NORTH AMERICA INC Syrris Atlas HD automated jacketed reactor with a modular platform offers interchangeability of reactors from 50mL to 5L
SDP
Supplier:  AGI NORTH AMERICA INC 20001ORBF
Encompass
Atlas HD was designed to facilitate safe walk-away chemistry to improve workflow.Included:-Touchscreen base for reaction automation-Data logging & real-time graphical display of reaction parameters-1L jacketed glass vessel (2L and 5L also available)-Quick-release clamp for rapid vessel change-Auto-aligning stirrer: 800 rpm and 11 Ncm torque-Stirrer seal & guide -PTFE 4-blade propeller stirrer-Oil drain unit to quickly remove thermal oil-Operating temperature range: -40 to 200 deg C (high range of -90 to 250 deg C available with upgrade kit)-Pressure range: Vacuum: ≤50 mbara. Maximum pressure: 250 mbar -Temperature probe and node-Pair of short oil pipes and connectors for reaction vessel to oil drain unitOrder informationNot included: appropriate circulator and oil pipes from the oil drain unit to the circulator. Oil pipes required M16 x 1.0 female connection and adapter depending on the circulator.
Catalog No. NC2417574
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.