Learn More
AGI NORTH AMERICA INC Syrris Orb Pilot floor-standing jacketed reactor for scale-up processes, available from 10L to 50L, equipped with 100Ncm stirrer motor
SDP
Supplier:  AGI NORTH AMERICA INC 30001OPBF
Encompass
Orb Pilot has multiple setups in one footprint.Included:-10L jacketed glass vessel (10 - 50L available separately)-Optimised jacket inlet to ensure high-efficiency heat transfer-Detachable outlet valve for easy cleaning-Base frame adaptable to multiple vessel sizes-Spring-loaded vessel support for easy vessel changes-Force-limited vessel clamp to prevent breakage from over-tightening-Operating temperature range: -40 to 235 deg C-Pressure range: Vacuum: ≤50 mbara. Maximum pressure: 250 mbar-LEMO RTD probe-100 Ncm stirrer motor for most demanding applications-Oil drain valve allows for easy draining of vessel jackets -Frame-mounted pair of short oil pipes with connectors-PTFE anchor stirrer with stirrer seal & guide -Lid blanking kitOrder informationNot included: appropriate circulator and pair of oil pipes from the frame-mounted short oil pipes to the circulator. Oil pipes required M30 x 1.5 female connection and adapter depending on the circulator.
Catalog No. NC2417578
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.