Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Orbiter Planetarium
SCI_ED

Catalog No. S45166 Shop All American Educational Products LLC Products
Change view
Click to view available options
Product Type:
Illuminated Orbiter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S45166 Illuminated Orbiter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S45166 Supplier American Educational Products Supplier No. 151

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lesson plan

Explain the cycles of day and night, the seasons, the phases of the moon and many other astronomy concepts.

  • Hand-operated gear-and-chain drive rotates the Earth on its axis, orients the axis in relation to the sun and revolves the Earth around the sun
  • Features a 6 in. sun (15.2cm), a 15 in. arm (38.1cm), a 4 in. geophysical Earth globe (10.2cm), a cratered moon and a transparent hemisphere screen

Specifications

Grade 3 to 12
For Use With (Application) Explains the cycles of day and night, the seasons, the phases of the moon and many other astronomy concepts
Includes Lesson plan, 6 in. sun, 15 in. arm, 4 in. geophysical earth globe, cratered moon and transparent hemisphere screen
Product Type Illuminated Orbiter
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.