Learn More
MilliporeSigma™ Supelco™ ORBO™ 2000 Precleaned Large PUF Cartridge
Catalog No. 111004477 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
6 cm
Catalog No. Diameter (Metric)
11-100-4477 6 cm
1 options
Catalog No. 11-100-4477 Supplier MilliporeSigma™ Supelco™ Supplier No. 20037
Add to Cart
Add to Cart
Used for gas and vapor sampling
ORBO™ 2000 precleaned large polyurethane foam (PUF) cartridge exhibit a low-pressure drop across the cartridge that facilitates high volume sampling. PUF adsorbent cartridges are useful for ASTM D6209, EPA IP-7 and EPA TO-13 (PAHs); EPA TO-4A (Pesticides/PCBs); EPA TO-9A (Dioxins) methods in monitoring semivolatiles in stack, ambient, indoor and workplace atmospheres.
TRUSTED_SUSTAINABILITY
Specifications
Description Assembled
Quantity 1/Pk.
Length (Metric) 7.6 cm
Diameter (Metric) 6 cm
ORBO is a trademark of Sigma-Aldrich Co. LLC