Learn More
MilliporeSigma™ Supelco™ ORBO™ 43 Supelpak-20 specially treated Amberlite XAD-2 (20/40), 100/50 mg W,W,W (specially treated) separators
Catalog No. 111010045 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
8 mm
Catalog No. Diameter (Metric)
11-101-0045 8 mm
1 options
Catalog No. 11-101-0045 Supplier MilliporeSigma™ Supelco™ Supplier No. 20258
Add to Cart
Add to Cart
Used for gas and vapor sampling
ORBO™ sorbent tubes contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed A 100 mg/Bed B 50 mg
Material W,W,W (Specially Treated) Separators
Pore Size 20 to 40 Mesh
Quantity 50/Pk.
Length (Metric) 100 mm
Diameter (Metric) 8 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC