Learn More
MilliporeSigma™ Supelco™ ORBO™ 49P (OVS) Supelpak-20 specially treated Amberlite XAD-2 (20/40), 270/140 mg RC,GFF,F,F (specially cleaned) separators
Catalog No. 111009937 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
13-8 mm
Catalog No. Diameter (Metric)
11-100-9937 13-8 mm
1 options
Catalog No. 11-100-9937 Supplier MilliporeSigma™ Supelco™ Supplier No. 20350
Add to Cart
Add to Cart
Used for gas and vapor sampling
The OSHA Versatile Sampler (OVS) was primarily designed for collection of Organophosphorus pesticides but is suitable for other applications. This specially constructed open glass tube is 75 mm long with a 13-mm outer diameter (O.D.) on the inlet end tapered to 8 mm (O.D.) on the outlet end. The design features a glass filter to trap the aerosols and a two-section sorbent portion to adsorb the vapors. The tube is packed with specially cleaned XAD-2 material and is QA tested to ensure low background interferences. The product is capped at the inlet and outlet to minimize environmental exposure. Meets OSHA and NIOSH method design specifications.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed A 270 mg/Bed B 140 mg
Material RC,GFF,F,F (Specially Cleaned) Separators
Pore Size 20 to 40 Mesh
Quantity 10/Pk.
Length (Metric) 75 mm
Diameter (Metric) 13-8 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC