Learn More
MilliporeSigma™ Supelco™ ORBO™ 615 Amberlite XAD-7 (15/50), 60/30 mg W,W,W separators
Catalog No. 111009879 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
6 mm
Catalog No. Diameter (Metric)
11-100-9879 6 mm
1 options
Catalog No. 11-100-9879 Supplier MilliporeSigma™ Supelco™ Supplier No. 20053
Add to Cart
Add to Cart
Used for gas and vapor sampling
ORBO™ 615 Amberlite™ XAD™-7 (15/50), 60/30 mg solvent desorption tube is designed for gas and vapor sampling.. It contains two beds of the same selective adsorbent separated by glass wool or foam.. Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed A 60 mg/Bed B 30 mg
Material W,W,W Separators
Pore Size 15-50 mesh
Quantity 50/Pk.
Length (Metric) 75 mm
Diameter (Metric) 6 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC