Learn More
Cayman Chemical IonmycIncalcIum salt 1mg
SDP
Supplier:  Cayman Chemical 119321 MG
Encompass_Preferred
Reversible inhibitor of JNK1 2 and 3 (IC50 0 11 M) is cell-permeable and inhibits c-Jun phosphorylation in cells blocks the expression of COX-2 and TNF-a in monocytes and IL-10 TNF-a and IFN-Y in T-cells prevents apoptosis in B-cells inhibits autophagy in HeLa cells
Catalog No. 50-295-3182
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.