Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582977 Supplier Invitrogen™ Supplier No. PA582977
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YISGVPGTGK TATVHEVIRC LQQAAQANDV PPFQYIEVNG MKLTEPHQVY VQILQKLTGQ KATANHAAEL LAKQFCTRGS PQETTVLLVD E Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 93%.

The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is the largest subunit of the ORC complex. While other ORC subunits are stable throughout the cell cycle, the levels of this protein vary during the cell cycle, which has been shown to be controlled by ubiquitin-mediated proteolysis after initiation of DNA replication. This protein is found to be selectively phosphorylated during mitosis. It is also reported to interact with MYST histone acetyltransferase 2 (MyST2/HBO1), a protein involved in control of transcription silencing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Orc1
Gene Accession No. Q13415
Gene Alias AA545195; HS; HSORC1; MmORC1; orc1; orc1.L; Orc1l; origin recognition complex protein 1; origin recognition complex subunit 1; origin recognition complex subunit 1 L homeolog; origin recognition complex, subunit 1; origin recognition complex, subunit 1 homolog; origin recognition complex, subunit 1-like; PARC1; Replication control protein 1; XELAEV_18023178mg; xorc1
Gene Symbols Orc1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ORC1. Recombinant protein control fragment (Product #RP-94042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.