Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561247
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1247 Supplier Bethyl Laboratories, Inc Supplier No. A301892A
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Origin recognition complex subunit 1 (ORC1L) is a member of the highly conserved six subunits of the Origin Recognition Complex (ORC) which is essential for the initiation of DNA replication in eukaryotic cells. ORC complex binds specifically to origins of replication and serves as a platform for further assembly of additional transcription initiation factors such as Cdc6 and Mcm proteins. ORC1L is the largest encoded protein of the complex. Levels of this protein vary during the cell cycle, which has been shown to be controlled by Ubiquitin-mediated proteolysis after initiation of DNA replication. This protein is found to be selectively phosphorylated during mitosis. It is also reported to interact with MYST histone acetyltransferase 2 (MyST2/HBO1), a protein involved in control of transcription silencing.

Specifications

Antigen ORC1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ORC1
Gene Accession No. Q13415
Gene Alias HSORC1, ORC1, PARC1
Gene Symbols ORC1
Host Species Rabbit
Immunogen Between 811 and 861
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4998
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.