Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ORC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584901 Supplier Invitrogen™ Supplier No. PA584901
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLCKQLEKIG QQVDREPGDV ATPPRKRKKI VVEAPAKEME KVEEMPHKPQ KDEDLTQDYE EWKRKILENA ASAQ Highest antigen sequence indentity to the following orthologs: Rat - 54%, Mouse - 61%.

The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Gene silencing studies with small interfering RNA demonstrated that this protein plays an essential role in coordinating chromosome replication and segregation with cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Orc6
Gene Accession No. Q9Y5N6
Gene Alias 6720420I10Rik; ORC 6; ORC6; ORC6L; origin recognition complex subunit 6; origin recognition complex, subunit 6; origin recognition complex, subunit 6-like
Gene Symbols Orc6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ORC6. Recombinant protein control fragment (Product #RP-105567).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23594
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.