Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Orexin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-459 Supplier Invitrogen™ Supplier No. PA19583
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9583 detects Orexin A in human, mouse, and rat samples. PA1-9583 has been successfully used in ELISA procedures. PA1-9583 immunogen is a synthetic peptide corresponding to amino acids 1-13 (AQPLPDCCRQKTC) of human Orexin A.

This gene encodes a hypothalamic neuropeptide precursor protein that gives rise to two mature neuropeptides, orexin A and orexin B, by proteolytic processing. Orexin A and orexin B, which bind to orphan G-protein coupled receptors HCRTR1 and HCRTR2, function in the regulation of sleep and arousal. This neuropeptide arrangement may also play a role in feeding behavior, metabolism, and homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin A
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HCRT
Gene Accession No. O43612, O55232, O55241
Gene Alias HCRT; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; OX; PPORX; PPOX; prepro-orexin
Gene Symbols HCRT
Host Species Chicken
Immunogen Synthetic peptide corresponding to amino acids 1-13 of human Orexin A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15171, 25723, 3060
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.